1,1'-(4,4'-Dimethoxy-[1,1'-biphenyl]-3,3'-diyl)bis(adamantane) ≥95%

Supplier: AOBChem USA

Warning

282847-1G 282847-500MG
77634-608EA 3417 USD
77634-608 77645-700
1,1'-(4,4'-Dimethoxy-[1,1'-biphenyl]-3,3'-diyl)bis(adamantane) ≥95%
1,1'-(4,4'-Dimethoxy-[1,1'-biphenyl]-3,3'-diyl)bis(adamantane)

Precaución: For Research Use Only

Formula: C₃₄H₄₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26142856
CAS Number: 932033-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR