1-(2-Bromophenoxy)-2-chloro-4-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

282786-5G 282786-1G 282786-25G 282786-500MG
77634-604EA 610 USD
77634-604 77637-742 77658-434 77649-778
1-(2-Bromophenoxy)-2-chloro-4-nitrobenzene ≥97%
1-(2-Bromophenoxy)-2-chloro-4-nitrobenzene

Precaución: For Research Use Only

Formula: C₁₂H₇BrClNO₃
Storage Temperature: Ambient
MDL Number: MFCD11124188
CAS Number: 1036437-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR