N,N-Diethylcyclohexanecarboxamide ≥97%

Supplier: AOBChem USA

Warning

282697-5G 282697-1G 282697-25G 282697-500MG
77634-600EA 428 USD
77634-600 77637-740 77658-430 77645-698
N,N-Diethylcyclohexanecarboxamide ≥97%
N,N-Diethylcyclohexanecarboxamide

Precaución: For Research Use Only

Formula: C₁₁H₂₁NO
Storage Temperature: Ambient
MDL Number: MFCD03374855
CAS Number: 5461-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR