B-[4-[(Cyclopropylcarbonyl)amino]phenyl]boronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
282555-5G
282555-1G
282555-500MG
282555-25G
77632-442EA
1206
USD
77632-442
77637-738
77652-584
77654-678
B-[4-[(Cyclopropylcarbonyl)amino]phenyl]boronic acid ≥95%
B-[4-[(Cyclopropylcarbonyl)amino]phenyl]boronic acid
Precaución: For Research Use Only
Formula:
205.02 g/mol MW: C₁₀H₁₂BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18383824 CAS Number: 1219080-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...