N-(3-Bromophenyl)cyclopropanecarboxamide ≥95%

Supplier: AOBChem USA

Warning

282546-5G 282546-1G 282546-25G 282546-500MG
77634-598EA 468 USD
77634-598 77637-736 77658-428 77645-696
N-(3-Bromophenyl)cyclopropanecarboxamide ≥95%
N-(3-Bromophenyl)cyclopropanecarboxamide

Precaución: For Research Use Only

Formula: 240.1 g/mol
MW: C₁₀H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD00029065
CAS Number: 14372-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR