N-Methylcyclopropanecarboxamide ≥95%

Supplier: AOBChem USA

Warning

282515-5G 282515-25G 282515-1G 282515-500MG
77632-440EA 618 USD
77632-440 77654-676 77641-708 77645-694
N-Methylcyclopropanecarboxamide ≥95%
N-Methylcyclopropanecarboxamide

Precaución: For Research Use Only

Formula: C₅H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00226845
CAS Number: 7108-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR