1-(2-Bromo-4,6-difluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

281590-5G 281590-1G 281590-500MG
77628-402EA 1484.23 USD
77628-402 77637-734 77652-582
1-(2-Bromo-4,6-difluorophenyl)ethanone ≥97%
1-(2-Bromo-4,6-difluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD28737867
CAS Number: 1805119-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR