Ethyl 3,5-di-tert-butyl-2-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

281485-5G 281485-25G 281485-1G 281485-500MG
77628-400EA 1484.23 USD
77628-400 77635-596 77641-704 77645-692
Ethyl 3,5-di-tert-butyl-2-ethoxybenzoate ≥97%
Ethyl 3,5-di-tert-butyl-2-ethoxybenzoate

Precaución: For Research Use Only

Formula: C₁₉H₃₀O₃
Storage Temperature: Ambient
MDL Number: MFCD34883638
CAS Number: 2969445-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR