Methyl 3,5-di-tert-butyl-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

281482-5G 281482-25G 281482-1G 281482-500MG
77628-398EA 1484.23 USD
77628-398 77635-594 77641-702 77645-690
Methyl 3,5-di-tert-butyl-2-methoxybenzoate ≥97%
Methyl 3,5-di-tert-butyl-2-methoxybenzoate

Precaución: For Research Use Only

Formula: 278.3865 g/mol
MW: C₁₇H₂₆O₃
Storage Temperature: Ambient
MDL Number: MFCD06203922
CAS Number: 195318-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR