3-Chloro-2,6-difluoroaniline ≥95%

Supplier: AOBChem USA

Warning

280858-5G 280858-25G
77634-596EA 106 USD
77634-596 77654-674
3-Chloro-2,6-difluoroaniline ≥95%
3-Chloro-2,6-difluoroaniline

Precaución: For Research Use Only

Formula: C₆H₄ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD06660173
CAS Number: 287172-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR