4-(3-Bromophenoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

280552-5G 280552-1G 280552-25G 280552-500MG
77632-436EA 902 USD
77632-436 77637-732 77654-672 77649-770
4-(3-Bromophenoxy)benzoic acid ≥95%
4-(3-Bromophenoxy)benzoic acid

Precaución: For Research Use Only

Formula: 293.113 g/mol
MW: C₁₃H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11156620
CAS Number: 347173-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR