2-(4-Methoxy-2-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

279698-1G 279698-5G 279698-500MG
77637-730EA 630 USD
77637-730 77654-670 77645-686
2-(4-Methoxy-2-methylphenyl)propan-2-ol ≥95%
2-(4-Methoxy-2-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD20528507
CAS Number: 91968-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR