1-(4-(Ethylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

279641-5G 279641-1G 279641-25G 279641-500MG
77628-394EA 1484.23 USD
77628-394 77637-726 77658-422 77645-684
1-(4-(Ethylthio)phenyl)cyclopentanol ≥95%
1-(4-(Ethylthio)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 222.3464 g/mol
MW: C₁₃H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34882600
CAS Number: 2969258-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR