2-Methoxy-3,5-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

27957-5G 27957-1G 27957-500MG 27957-25G
77632-432EA 885 USD
77632-432 77637-724 77652-578 77658-420
2-Methoxy-3,5-dimethylbenzoic acid ≥95%
2-Methoxy-3,5-dimethylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD14537327
CAS Number: 21553-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR