2-Bromo-1-chloro-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

279155-5G 279155-25G 279155-1G 279155-500MG
77632-430EA 912 USD
77632-430 77658-418 77641-696 77645-680
2-Bromo-1-chloro-3-fluoro-4-methoxybenzene ≥97%
2-Bromo-1-chloro-3-fluoro-4-methoxybenzene

Precaución: For Research Use Only

Formula: 239.469 g/mol
MW: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD29043821
CAS Number: 1785595-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR