1-(5-Methyl-3-thienyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

278938-5G 278938-1G 278938-500MG
77628-392EA 1484.23 USD
77628-392 77641-694 77645-678
1-(5-Methyl-3-thienyl)-1-propanone ≥95%
1-(5-Methyl-3-thienyl)-1-propanone

Precaución: For Research Use Only

Formula: 154.23 g/mol
MW: C₈H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD21125934
CAS Number: 106265-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR