3,5-Dimethyl-2-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

278898-5G 278898-25G 278898-1G 278898-500MG
77628-390EA 1484.23 USD
77628-390 77658-416 77641-692 77645-676
3,5-Dimethyl-2-(methylthio)thiophene ≥95%
3,5-Dimethyl-2-(methylthio)thiophene

Precaución: For Research Use Only

Formula: 158.29 g/mol
MW: C₇H₁₀S₂
Storage Temperature: Ambient
MDL Number: MFCD34882061
CAS Number: 69442-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR