1-[3-(1,1-Dimethylethyl)-4-methoxyphenyl]-2-methyl-1-propanone ≥95%
Supplier: AOBChem USA
|
Warning
|
278875-5G
278875-25G
278875-1G
278875-500MG
77628-388EA
1484.23
USD
77628-388
77658-414
77641-690
77645-674
1-[3-(1,1-Dimethylethyl)-4-methoxyphenyl]-2-methyl-1-propanone ≥95%
1-[3-(1,1-Dimethylethyl)-4-methoxyphenyl]-2-methyl-1-propanone
Precaución: For Research Use Only
Formula:
234.33 g/mol MW: C₁₅H₂₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD14530820 CAS Number: 1216063-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...