Methanone, [3-(1,1-dimethylethyl)-4-methoxyphenyl]phenyl- ≥95%
Supplier: AOBChem USA
|
Warning
|
278869-5G
278869-25G
278869-1G
278869-500MG
77628-386EA
1484.23
USD
77628-386
77635-592
77637-720
77649-766
Methanone, [3-(1,1-dimethylethyl)-4-methoxyphenyl]phenyl- ≥95%
Methanone, [3-(1,1-dimethylethyl)-4-methoxyphenyl]phenyl-
Precaución: For Research Use Only
Formula:
268.35 g/mol MW: C₁₈H₂₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD34882057 CAS Number: 1536965-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...