3,5-Dimethyl-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27871-5G 27871-25G 27871-1G 27871-500MG
77634-594EA 536 USD
77634-594 77654-664 77641-688 77649-762
3,5-Dimethyl-4-iodobenzaldehyde ≥97%
3,5-Dimethyl-4-iodobenzaldehyde

Precaución: For Research Use Only

Formula: 260.072 g/mol
MW: C₉H₉IO
Storage Temperature: Ambient
MDL Number: MFCD27665651
CAS Number: 99357-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR