(5-Methoxy-2-methylphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

278672-5G 278672-25G 278672-1G 278672-500MG
77628-384EA 1484.23 USD
77628-384 77654-662 77641-686 77649-760
(5-Methoxy-2-methylphenyl)(phenyl)methanone ≥95%
(5-Methoxy-2-methylphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 226.27 g/mol
MW: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD22492743
CAS Number: 2734021-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR