[2-Methoxy-5-(trifluoromethyl)phenyl]phenylmethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
278637-5G
278637-1G
278637-25G
278637-500MG
77628-380EA
1484.23
USD
77628-380
77637-712
77654-660
77649-758
[2-Methoxy-5-(trifluoromethyl)phenyl]phenylmethanone ≥95%
[2-Methoxy-5-(trifluoromethyl)phenyl]phenylmethanone
Precaución: For Research Use Only
Formula:
280.24 g/mol MW: C₁₅H₁₁F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22492760 CAS Number: 72083-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...