(5-Chloro-2-methoxyphenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

278633-5G 278633-1G 278633-500MG
77658-410EA 2001 USD
77658-410 77645-666 77649-756
(5-Chloro-2-methoxyphenyl)phenylmethanone ≥95%
(5-Chloro-2-methoxyphenyl)phenylmethanone

Precaución: For Research Use Only

Formula: 246.69 g/mol
MW: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11210577
CAS Number: 4072-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR