2-Chloro-4-(ethylthio)-1-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

278599-5G 278599-25G 278599-1G 278599-500MG
77628-378EA 1484.23 USD
77628-378 77654-658 77641-684 77649-754
2-Chloro-4-(ethylthio)-1-(trifluoromethyl)benzene ≥95%
2-Chloro-4-(ethylthio)-1-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 240.67 g/mol
MW: C₉H₈ClF₃S
Storage Temperature: Ambient
MDL Number: MFCD22483689
CAS Number: 1822982-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR