1-(2-Chlorophenyl)-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

278405-5G 278405-25G 278405-1G 278405-500MG
77632-424EA 691 USD
77632-424 77654-656 77645-660 77645-662
1-(2-Chlorophenyl)-1H-pyrazole-4-carboxylic acid ≥97%
1-(2-Chlorophenyl)-1H-pyrazole-4-carboxylic acid

Precaución: For Research Use Only

Formula: 222.63 g/mol
MW: C₁₀H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12168893
CAS Number: 1154198-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR