1-[4-(Methylthio)phenyl]-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

278148-5G 278148-1G 278148-25G 278148-500MG
77628-376EA 1484.23 USD
77628-376 77637-706 77658-406 77645-656
1-[4-(Methylthio)phenyl]-1H-pyrazole ≥97%
1-[4-(Methylthio)phenyl]-1H-pyrazole

Precaución: For Research Use Only

Formula: 190.27 g/mol
MW: C₁₀H₁₀N₂S
Storage Temperature: Ambient
MDL Number: MFCD25964810
CAS Number: 572912-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR