2-Bromo-3-fluoro-4-methoxyacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
27766-5G
27766-500MG
27766-25G
27766-1G
77634-592EA
536
USD
77634-592
77652-572
77658-400
77641-680
2-Bromo-3-fluoro-4-methoxyacetophenone ≥97%
2-Bromo-3-fluoro-4-methoxyacetophenone
Precaución: For Research Use Only
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28142618 CAS Number: 1809161-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...