2-Methoxy-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

277540-5G 277540-25G 277540-1G 277540-500MG
77632-416EA 1288 USD
77632-416 77654-650 77645-650 77649-752
2-Methoxy-α-[2-(trimethylsilyl)ethynyl]benzenemethanol ≥95%
2-Methoxy-α-[2-(trimethylsilyl)ethynyl]benzenemethanol

Precaución: For Research Use Only

Formula: 234.37 g/mol
MW: C₁₃H₁₈O₂Si
Storage Temperature: Ambient
MDL Number: MFCD35169093
CAS Number: 153889-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR