5-Methyl-2-(methylthio)pyridine-3-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
27736-1G
27736-5G
27736-500MG
77637-704EA
563
USD
77637-704
77658-398
77645-648
5-Methyl-2-(methylthio)pyridine-3-boronic acid ≥97%
5-Methyl-2-(methylthio)pyridine-3-boronic acid
Precaución: For Research Use Only
Formula:
183.036 g/mol MW: C₇H₁₀BNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD28142616 CAS Number: 2121513-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...