1-(3-(Benzyloxy)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

27730-5G 27730-1G 27730-500MG
77628-372EA 1599.59 USD
77628-372 77637-702 77649-748
1-(3-(Benzyloxy)phenyl)cyclobutanol ≥95%
1-(3-(Benzyloxy)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₇H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD31613897
CAS Number: 211495-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR