4-Phenoxy-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27720-5G 27720-25G 27720-1G 27720-500MG
77628-370EA 1484.23 USD
77628-370 77635-588 77641-676 77649-746
4-Phenoxy-2-methoxyphenylboronic acid ≥97%
4-Phenoxy-2-methoxyphenylboronic acid

Precaución: For Research Use Only

Formula: 244.051 g/mol
MW: C₁₃H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD28977262
CAS Number: 2899272-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR