6-(Trifluoromethyl)quinolin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

276446-5G 276446-25G 276446-1G 276446-500MG
77632-414EA 747 USD
77632-414 77654-646 77645-644 77649-742
6-(Trifluoromethyl)quinolin-2(1H)-one ≥97%
6-(Trifluoromethyl)quinolin-2(1H)-one

Precaución: For Research Use Only

Formula: C₁₀H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD11108684
CAS Number: 835903-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR