Ethyl (2-chloro-4-fluoropyridin-3-yl)acetate ≥97%

Supplier: AOBChem USA

Warning

27634-5G 27634-1G 27634-500MG
77628-362EA 1571.78 USD
77628-362 77637-694 77649-738
Ethyl (2-chloro-4-fluoropyridin-3-yl)acetate ≥97%
Ethyl (2-chloro-4-fluoropyridin-3-yl)acetate

Precaución: For Research Use Only

Formula: 203.598 g/mol
MW: C₈H₇ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD28752437
CAS Number: 1807259-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR