5-Ethoxy-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

27598-5G 27598-1G 27598-500MG 27598-25G
77632-412EA 940 USD
77632-412 77637-692 77652-570 77654-642
5-Ethoxy-2-methylbenzoic acid ≥95%
5-Ethoxy-2-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20640165
CAS Number: 112868-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR