3-Fluoro-2-trifluoromethylisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-(trifluoromethyl)isonicotinic acid

275905-5G 275905-25G
77634-588EA 159 USD
77634-588 77658-396
3-Fluoro-2-trifluoromethylisonicotinic acid ≥97%
3-Fluoro-2-trifluoromethylisonicotinic acid

Precaución: For Research Use Only

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437597
CAS Number: 886510-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR