3-(3-Methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

27552-5G 27552-1G 27552-500MG
77628-360EA 1484.23 USD
77628-360 77637-690 77652-566
3-(3-Methylphenyl)pyridine ≥97%
3-(3-Methylphenyl)pyridine

Precaución: For Research Use Only

Formula: 169.222 g/mol
MW: C₁₂H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD06801823
CAS Number: 4385-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR