2-Bromo-4,6-dichlorobenzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

27543-5G 27543-25G 27543-1G 27543-500MG
77628-358EA 1484.23 USD
77628-358 77635-582 77641-672 77645-640
2-Bromo-4,6-dichlorobenzoic acid methyl ester ≥95%
2-Bromo-4,6-dichlorobenzoic acid methyl ester

Precaución: For Research Use Only

Formula: C₈H₅BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29088127
CAS Number: 943975-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR