2-Bromo-4,6-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27542-5G 27542-1G 27542-25G 27542-500MG
77632-410EA 1168 USD
77632-410 77637-688 77654-640 77649-736
2-Bromo-4,6-dichlorobenzoic acid ≥97%
2-Bromo-4,6-dichlorobenzoic acid

Precaución: For Research Use Only

Formula: C₇H₃BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27926964
CAS Number: 650598-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR