1-(4-Fluoro[1,1'-biphenyl]-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

275358-1G 275358-5G 275358-500MG
77637-686EA 657 USD
77637-686 77658-392 77649-734
1-(4-Fluoro[1,1'-biphenyl]-3-yl)ethanone ≥95%
1-(4-Fluoro[1,1'-biphenyl]-3-yl)ethanone

Precaución: For Research Use Only

Formula: 214.24 g/mol
MW: C₁₄H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD26687733
CAS Number: 1539639-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR