1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

275319-5G 275319-1G 275319-25G 275319-500MG
77628-354EA 1484.23 USD
77628-354 77637-684 77654-638 77649-730
1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanone ≥95%
1-(5-Bromo-3-ethyl-2-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD34881124
CAS Number: 2708390-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR