3-Bromo-4-methyl-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

27515-5G 27515-1G 27515-25G 27515-500MG
77632-408EA 749 USD
77632-408 77637-682 77658-388 77645-638
3-Bromo-4-methyl-5-(methylthio)pyridine ≥97%
3-Bromo-4-methyl-5-(methylthio)pyridine

Precaución: For Research Use Only

Formula: 218.114 g/mol
MW: C₇H₈BrNS
Storage Temperature: Ambient
MDL Number: MFCD30529799
CAS Number: 2056110-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR