3-Flouro-2-iodo-4-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

27496-5G 27496-25G 27496-1G 27496-500MG
77632-406EA 1117 USD
77632-406 77658-386 77641-668 77649-724
3-Flouro-2-iodo-4-(trimethylsilyl)pyridine ≥97%
3-Flouro-2-iodo-4-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: C₈H₁₁FINSi
Storage Temperature: Ambient
MDL Number: MFCD28142596
CAS Number: 1809161-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR