3-Fluoro-2-(thiomethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

27493-5G 27493-1G 27493-500MG 27493-25G
77632-404EA 700 USD
77632-404 77637-674 77652-562 77654-632
3-Fluoro-2-(thiomethyl)pyridine ≥95%
3-Fluoro-2-(thiomethyl)pyridine

Precaución: For Research Use Only

Formula: 143.182 g/mol
MW: C₆H₆FNS
Storage Temperature: Ambient
MDL Number: MFCD21365088
CAS Number: 902518-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR