3-Fluoro-4-(methylthio)-pyridine ≥97%

Supplier: AOBChem USA

Warning

27492-5G 27492-25G
77634-584EA 98 USD
77634-584 77658-384
3-Fluoro-4-(methylthio)-pyridine ≥97%
3-Fluoro-4-(methylthio)-pyridine

Precaución: For Research Use Only

Formula: C₆H₆FNS
MW: 143.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142593
CAS Number: 902518-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR