α-(2,3-Difluorophenyl)-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

274313-5G 274313-1G 274313-500MG
77628-350EA 1484.23 USD
77628-350 77637-672 77649-722
α-(2,3-Difluorophenyl)-2-pyridinemethanol ≥95%
α-(2,3-Difluorophenyl)-2-pyridinemethanol

Precaución: For Research Use Only

Formula: 221.2 g/mol
MW: C₁₂H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD19626912
CAS Number: 1339381-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR