α-(2,5-Difluorophenyl)-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

274258-5G 274258-25G 274258-1G 274258-500MG
77632-402EA 1381 USD
77632-402 77654-630 77641-666 77645-636
α-(2,5-Difluorophenyl)-2-pyridinemethanol ≥95%
α-(2,5-Difluorophenyl)-2-pyridinemethanol

Precaución: For Research Use Only

Formula: 221.2 g/mol
MW: C₁₂H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD17223862
CAS Number: 558465-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR