α-(2-Chlorophenyl)-2-pyridinemethanol ≥97%
Supplier: AOBChem USA
|
Warning
|
274202-5G
274202-25G
274202-1G
274202-500MG
77632-400EA
1206
USD
77632-400
77654-628
77641-664
77649-720
α-(2-Chlorophenyl)-2-pyridinemethanol ≥97%
α-(2-Chlorophenyl)-2-pyridinemethanol
Precaución: For Research Use Only
Formula:
C₁₂H₁₀ClNO Storage Temperature: Ambient |
MDL Number:
MFCD12824579 CAS Number: 6238-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...