α-(3-Methylphenyl)-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

274199-5G 274199-25G 274199-1G 274199-500MG
77632-398EA 1206 USD
77632-398 77654-626 77641-662 77649-718
α-(3-Methylphenyl)-2-pyridinemethanol ≥95%
α-(3-Methylphenyl)-2-pyridinemethanol

Precaución: For Research Use Only

Formula: 199.25 g/mol
MW: C₁₃H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD12153523
CAS Number: 29263-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR