2,6-Dichloro-5-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

274180-1G 274180-5G 274180-500MG
77634-582EA 1003 USD
77634-582 77654-624 77645-634
2,6-Dichloro-5-methylnicotinaldehyde ≥95%
2,6-Dichloro-5-methylnicotinaldehyde

Precaución: For Research Use Only

Formula: C₇H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD34167977
CAS Number: 2386302-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR