4-Cyano-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

27408-5G 27408-25G 27408-1G 27408-500MG
77632-394EA 1033 USD
77632-394 77658-380 77641-660 77649-714
4-Cyano-3-iodopyridine ≥97%
4-Cyano-3-iodopyridine

Precaución: For Research Use Only

Formula: C₆H₃IN₂
MW: 230.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08729285
CAS Number: 10386-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR